Få mere at vide
(RS)-N-(2,6-Dimethylphenyl)piperidine-2-carboxamide, Mikromol™
Artikelnummer. 30051635
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30051635 25 mg
17902356 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30051635 Leverandør Mikromol Leverandørnr. MM0368.080025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(RS)-N-(2,6-Dimethylphenyl)piperidine-2-carboxamide
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (RS)-N-(2,6-Dimethylphenyl)piperidine-2-carboxamide
CAS 15883-20-2
Analyt- eller komponentnavne (RS)-N-(2,6-Dimethylphenyl)piperidine-2-carboxamide
Forsendelsestilstand Room Temperature
Molekylær formel C14 H20 N2 O
Certificeringer/compliance ISO 17025
Synonym (RS)-N-(2,6-Dimethylphenyl)piperidine-2-carboxamide, Mepivacaine Imp. B (EP), Bupivacaine Imp. B (EP), Bupivacaine Hydrochloride Imp. B (EP), Mepivacaine Hydrochloride Imp. B (EP), Bupivacaine USP Related Compound B, Bupivacaine USP RC B, Bupivacaine Related Compound B, Bupivacaine Hydrochloride Impurity B, Mepivacaine Hydrochloride Impurity B, Bupivacaine Impurity B, Mepivacaine Impurity B
InChI formel InChI=1S/C14H20N2O/c1-10-6-5-7-11(2)13(10)16-14(17)12-8-3-4-9-15-12/h5-7,12,15H,3-4,8-9H2,1-2H3,(H,16,17)
SMIL Cc1cccc(C)c1NC(=O)C2CCCCN2
IUPAC navn N-(2,6-dimethylphenyl)piperidine-2-carboxamide
Molekylvægt (g/mol) 232.32
Mængde 25 mg
Formel vægt 232.1576 g/mol