Få mere at vide
(RS)-N-(2,6-Dimethylphenyl)piperidine-2-carboxamide Hydrochloride, Mikromol™
Beskrivelse
(RS)-N-(2,6-Dimethylphenyl)piperidine-2-carboxamide Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (2RS)-N-(2,6-dimethylphenyl)piperidine-2-carboxamide |
| CAS | 65797-42-4 |
| Analyt- eller komponentnavne | (2RS)-N-(2,6-dimethylphenyl)piperidine-2-carboxamide |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C14 H20 N2 O . Cl H |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Mepivacaine Hydrochloride Imp. B (EP), Mepivacaine Imp. B (EP), Bupivacaine USP Related Compound B, Bupivacaine USP RC B, (RS)-N-(2,6-Dimethylphenyl)piperidine-2-carboxamide Hydrochloride, Mepivacaine Hydrochloride Imp. B (EP) as Hydrochloride, Bupivacaine Related Compound B as Hydrochloride, Bupivacaine Hydrochloride Impurity B as Hydrochloride, Mepivacaine Hydrochloride Impurity B as Hydrochloride, Bupivacaine Impurity B as Hydrochloride, Mepivacaine Impurity B as Hydrochloride |
| InChI formel | InChI=1S/C14H20N2O.ClH/c1-10-6-5-7-11(2)13(10)16-14(17)12-8-3-4-9-15-12;/h5-7,12,15H,3-4,8-9H2,1-2H3,(H,16,17);1H |
| SMIL | Cl.Cc1cccc(C)c1NC(=O)C2CCCCN2 |
| IUPAC navn | N-(2,6-dimethylphenyl)piperidine-2-carboxamide;hydrochloride |
| Vis mere |