Få mere at vide
(RS)-9,10-Difluoro-3-methyl-7-oxo-2,3-dihydro-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic Acid (FPA), Mikromol™
Artikelnummer. 30051334
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30051334 25 mg
30050176 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30051334 Leverandør Mikromol Leverandørnr. MM0228.040025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(RS)-9,10-Difluoro-3-methyl-7-oxo-2,3-dihydro-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic Acid (FPA)
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (RS)-9,10-difluoro-3-methyl-7-oxo-2,3-dihydro-7H-pyrido [1,2,3-de][1,4]benzoxazine-6-carboxylic acid (FPA)
CAS 82419-35-0
Analyt- eller komponentnavne (RS)-9,10-difluoro-3-methyl-7-oxo-2,3-dihydro-7H-pyrido [1,2,3-de][1,4]benzoxazine-6-carboxylic acid (FPA)
Forsendelsestilstand Room Temperature
Molekylær formel C13 H9 F2 N O4
Certificeringer/compliance ISO 9001
Synonym (RS)-9,10-Difluoro-3-methyl-7-oxo-2,3-dihydro-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic Acid, Ofloxacin Imp. A (EP), FPA, Ofloxacin Impurity A
InChI formel InChI=1S/C13H9F2NO4/c1-5-4-20-12-9(15)8(14)2-6-10(12)16(5)3-7(11(6)17)13(18)19/h2-3,5H,4H2,1H3,(H,18,19)
SMIL CC1COc2c(F)c(F)cc3C(=O)C(=CN1c23)C(=O)O
IUPAC navn (RS)-9,10-Difluoro-3-methyl-7-oxo-2,3-dihydro-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid
Molekylvægt (g/mol) 281.21
Mængde 25 mg
Formel vægt 281.05 g/mol