Få mere at vide
(RS)-4-(Aminomethyl)cyclohex-1-enecarboxylic Acid Hydrochloride, Mikromol™
Beskrivelse
(RS)-4-(Aminomethyl)cyclohex-1-enecarboxylic Acid Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (RS)-4-(Aminomethyl)cyclohex-1-enecarboxylic Acid Hydrochloride |
| CAS | 1803601-44-6 |
| Analyt- eller komponentnavne | (RS)-4-(Aminomethyl)cyclohex-1-enecarboxylic Acid Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C8 H13 N O2 . Cl H |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Tranexamic Acid Imp. C (EP), Tranexamic Acid USP Related Compound C, Tranexamic Acid USP RC C, (RS)-4-(Aminomethyl)cyclohex-1-enecarboxylic Acid Hydrochloride, Tranexamic Acid Related Compound C as Hydrochloride, Tranexamic Acid Impurity C as Hydrochloride |
| InChI formel | InChI=1S/C14H21N3O.2ClH/c1-17-10-5-8-13(9-11-17)15-16-14(18)12-6-3-2-4-7-12;;/h2-4,6-7,13,15H,5,8-11H2,1H3,(H,16,18);2*1H |
| SMIL | Cl.NCC1CCC(=CC1)C(=O)O |
| IUPAC navn | 4-(aminomethyl)cyclohexene-1-carboxylic acid;hydrochloride |
| Vis mere |