Få mere at vide
(RS)-3,5-DHPG, TRC
Artikelnummer. 30378768
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30378768 10 mg
30380478 100 mg
30384432 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30378768 Supplier Toronto Research Chemicals Supplier No. TRCD45002810MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 146255-66-5
Molekylær formel C8H9NO4
Molekylvægt (g/mol) 183.16
Synonym alpha-Amino-3,5-dihydroxybenzeneacetic Acid, (RS)-3,5-Dihydroxyphenylglycine, (±)-alpha-Amino-3,5-dihydroxybenzeneacetic Acid, 3,5-Dihydroxyphenylglycine, DHPG, DL-2-(3,5-Dihydroxyphenyl)glycine, (D/L)-3,5-Dihydroxyphenylglycine, 2-Amino-2-(3,5-dihydroxyphenyl)acetic Acid
SMIL NC(C(=O)O)c1cc(O)cc(O)c1
Specifications
Kemisk navn eller materiale (RS)-3,5-DHPG
Mængde 10 mg
InChI formel InChI=1S/C8H9NO4/c9-7(8(12)13)4-1-5(10)3-6(11)2-4/h1-3,7,10-11H,9H2,(H,12,13)
Formel vægt 183.0532