Få mere at vide
(RS)-2-[(2-Methylphenyl)phenylmethoxy]ethanamine Hydrochloride, Mikromol™
Beskrivelse
(RS)-2-[(2-Methylphenyl)phenylmethoxy]ethanamine Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (RS)-2-[(2-Methylphenyl)phenylmethoxy]ethanamine Hydrochloride |
| CAS | 17752-32-8 |
| Analyt- eller komponentnavne | (RS)-2-[(2-Methylphenyl)phenylmethoxy]ethanamine Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C16 H19 N O . Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Orphenadrine Hydrochloride Imp. C (EP), Orphenadrine Imp. C (EP), (RS)-2-[(2-Methylphenyl)phenylmethoxy]ethanamine Hydrochloride, Orphenadrine Hydrochloride Imp. C (EP) as Hydrochloride, Ethanamine, 2-[(2-methylphenyl)phenylmethoxy]-, hydrochloride (1:1), Ethanamine, 2-[(2-methylphenyl)phenylmethoxy]-, hydrochloride (9CI), Ethylamine, 2-[(o-methyl-alpha-phenylbenzyl)oxy]-, hydrochloride (8CI), (o-Methyl-alpha-phenylbenzyloxy)ethylamine, hydrochloride, N,N-Didemethylorphenadrine hydrochloride, Orphenadrine Hydrochloride Impurity C as Hydrochloride, Orphenadrine Impurity C as Hydrochloride |
| InChI formel | InChI=1S/C16H19NO.ClH/c1-13-7-5-6-10-15(13)16(18-12-11-17)14-8-3-2-4-9-14;/h2-10,16H,11-12,17H2,1H3;1H |
| SMIL | Cl.Cc1ccccc1C(OCCN)c2ccccc2 |
| IUPAC navn | 2-[(2-methylphenyl)-phenylmethoxy]ethanamine;hydrochloride |
| Vis mere |