missing translation for 'onlineSavingsMsg'
Få mere at vide

Tocris Bioscience™ RS 100329 hydrochloride
GREENER_CHOICE

Artikelnummer. 18064412
Change view
Click to view available options
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
18064412 50 mg
18045072 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18064412 Supplier Tocris Bioscience™ Supplier No. 1325/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent, subtype-selective α1A antagonist

Subtype-selective α1A-adrenoceptor antagonist (pKi = 9.6 for human cloned α1A receptors). Displays 126- and 50-fold selectivity over human α1B and α1D receptors respectively. Active in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1215654-26-4
Molekylær formel C20H26ClF3N4O3

Specifications

Kemisk navn eller materiale RS 100329 hydrochloride
Mængde 50 mg
Formel vægt 462.89
Renhed >99%
Molekylvægt (g/mol) 462.898
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.