missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human RPS6KB1 (p70S6K), GST Tag Recombinant Protein
Human RPS6KB1 (p70S6K), GST Tag Recombinant Protein
4375.00 DKK
Specifications
Accession Number | NP_003152,T412E |
---|---|
For Use With (Application) | Kinase Assay |
Molecular Weight (g/mol) | 75.1 kDa |
Gene Alias | RPS6KB1 (p70S6K) |
KinaseFamily | RSK (RPS6K) Kinase Family |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
11583547
|
Invitrogen
PV3815 |
10μg |
4375.00 DKK
10µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
RPS6KB1 (p70S6K) is a serine⁄threonine kinase with 2 non-identical kinase catalytic domains which phosphorylates several residues of the S6 ribosomal protein.Specifications
NP_003152,T412E | |
75.1 kDa | |
RSK (RPS6K) Kinase Family | |
Recombinant | |
Storage Buffer: 50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice |
Kinase Assay | |
RPS6KB1 (p70S6K) | |
Human | |
GST Tag | |
Ser⁄Thr Kinases: AGC Group | |
Baculovirus |