missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
RPS27L Rabbit anti-Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-85672-0.1ML
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
RPS27L Polyclonal specifically detects RPS27L in Rat samples. It is validated for Western Blot.
Tekniske data
| RPS27L | |
| Polyclonal | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 51065 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| 40S ribosomal protein S27-like, ribosomal protein S27-like | |
| The immunogen is a synthetic peptide directed towards the middle region of Rat RPS27L | |
| 0.1 mL | |
| Primary | |
| Rat | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion