Learn More
Lonza™ RPMI 1640 with UltraGlutaminetm, 500mL
Developed for culturing normal human leukocytes. Lonza™ RPMI 1640 with UltraGlutaminetm, 500mL can be used for suspension and monolayer mammalian cultures.
Brand:  Lonza™ BE12-702F
Additional Details : Weight : 0.64000kg
Product Code. 11685210
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Developed for culturing normal human leukocytes
  • Can be used for suspension and monolayer mammalian cultures
  • Will work with different cell lines
  • Can also be used with primary cells such as blood/hematopoietc cells
Specifications
Specifications
Cell Culture Media
2 g/L D-Glucose anhydrous, 2 g/L NaHCO3, 300mg/L L-Glutamine, 400mg/L KCl, 6 g/L NaCl, 100mg/L Ca(NO3)2.4H2O, 1mg/L Glutathione reduce, 241.86mg/L L-Arginine. HCl, 56.82mg/L L-Asparagine.H2O, 20mg/L L-Aspartic Acid, 10mg/L Glycine, 20mg/L L-Glutamic Acid,
500 mL