Learn More
Lenz Laborglasinstrumente™ DURAN™ Round Bottom Flasks
Can be used to make solutions or hold, contain and collect chemicals. Lenz Laborglasinstrumente™ DURAN™ Round Bottom Flasks are manufactured from durable DURAN glass.
Brand:  Lenz Laborglasinstrumente™ B0418211
Additional Details : Weight : 0.09900kg
Product Code. 10499190
  • 115.00 DKK / Each
Estimated Shipment: 11-06-2024
to see stock.
Add to basket
Description
Description
  • With inscription label
  • Corrosion-free and resistant to thermal shock
Specifications
Specifications
250 mL
Borosilicate Glass, Duran Glass
NS 29
Round