Få mere at vide
Korff™ Runde aluminiumsfolieskiver
Product Code. 10728521
Change view
Click to view available options
Diameter (metrisk) Udvendig:
100 mm
120 mm
130 mm
150 mm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (metrisk) Udvendig
10728521 150 mm
10313841 100 mm
12682603 100 mm
10272041 120 mm
12672603 130 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10728521 Supplier Korff™ Supplier No. 60220
Spotlight-tilbud
Brug rabatkode 28038
View all promotions
Forventet forsendelse: 30-12-2026
Specifications
Diameter (metrisk) Udvendig 150 mm
Materiale Aluminium
Til brug med (applikation) Anvendes til mange forskellige laboratorieopgaver