missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Rotor S-4x750
Medium capacity swing-bucket rotor with max. speed 4,800 x g (4,700 rpm); includes 4 x 750mL round buckets
11865.00 DKK - 25830.00 DKK
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
15758369
|
Eppendorf™
5895120008 |
Rotor S-4x750, incl. 4 × 750 mL round buckets |
25830.00 DKK
Each |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15768369
|
Eppendorf™
5895121004 |
Rotor S-4x750, without buckets |
11865.00 DKK
Each |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Broad range of adapters enables high versatility for tubes and flasks from 0.2mL to 750mL and plates
- Mixed loading with 2 plate buckets and 2 round tube buckets allows for increased versatility
- Aerosol-tight QuickLock caps for safe centrifugation of hazardous samples optionally available
- Buckets, caps and adapters are autoclavable