missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Rotor S-4x400
Rotor S-4x400 is a high-speed, medium capacity, swing-bucket rotor suitable for use with the 5910 R refrigerated centrifuge.
10920.00 DKK
Specifications
Capacity | 0.2 to 250 mL |
---|---|
Max. Speed | 5,100 rpm |
For Use With (Equipment) | 5910 R Centrifuge |
No. of Places | 4 |
Biocontainment | Aerosol-tight caps available |
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
15833212
|
Eppendorf™
5895181007 |
Without buckets |
10920.00 DKK
Each |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- High-speed medium capacity swing-bucket rotor
- Broad range of adapters enables high versatility for tubes and bottles
- Aerosol-tight QuickLock caps for safe centrifugation of hazardous samples optionally available
- Buckets, caps and adapters are autoclavable
- Requires buckets to accommodate tubes and bottles from 0.2 to 250 mL
Specifications
0.2 to 250 mL | |
5910 R Centrifuge | |
Aerosol-tight caps available |
5,100 rpm | |
4 |