Beskrivelse
- Instead of a jewel bearing, these devices are equipped with a special ball bearing technology.
- They are characterized by their high robustness. Predestined for applications with very high sample throughput, frequent spindle changes, operation in harsher environments, or even frequently changing device locations.
- Whether simple or demanding viscosity measurements - with ROTAVISC you get fast and accurate results.
- Easy to use
- Intuitive menu navigation awaits you via the generous 4.3'' TFT display. A digital leveling also supports the correct setting of the starting condition. Recurring tasks are facilitated by the simple ramp function.
- Highest measurement accuracy
- The accuracy of the ROTAVISC rotary viscometers for Newtonian and non-Newtonian liquids is ± 1 % of the measuring range. The reproducibility is ± 0.2%.
- Infinitely variable speed
- With ROTAVISC you have a viscometer in which you can adjust the speed stepless, but also specify programs or individual speed levels.
- IKA ROTAVISC SBS hi-vi I Complete
- Viscosity-Measuring range: 200 to 80,000,000 mPas
Tekniske data
Tekniske data
| Produkttype | Viskosimeter |
| Viskositetsområde | 200 to 80,000,000 mPa |
| Display Type | TFT-skærm |
| Nøjagtighed | ± 1% |
| Gentagelighed | 0.002 |
| Sensorer | PT 100 |
| Hastighedsområde | 0.01 to 200 rpm |
| Temperaturområde (metrisk) | -100 °C til +300 °C |
| Termometer opløsning | 0.1 K |
| Momentområde | 1.4374 mNm |
| Vis mere |