Få mere at vide
Fisherbrand™ Rotary Vane Vacuum Pump
Shop alle Fisherbrand produkter
Flowhastighed:
120 to 143 L/min.
30 to 36 L/min.
70 to 92 L/min.
97 to 110 L/min.
Pakningsstørrelse:
Hver
Beskrivelse
- Two-stage direct driven rotary vane pump
- Designed for wide range of physical and chemical laboratory applications and continuous running
- Low weight, small footprint and high pumping speed, increased chemical resistance are advantageous features in research and routine processes
- Rugged reliable design, combined with low noise level and ease of use
- Innovative gas ballast valve prevents moisture condensing and high tolerance of water and solvent vapors
- Intake line is vacuum sealed by and anti suckback valve to protect your application from undesired venting and oil back flow after pump is turned off
- Vertical oil sight glass allows easy monitoring of oil level and condition
Tekniske data
Tekniske data
| Maks. Vakuum | 2 x 10-3 mbar |
| Elektriske krav | 115/230 V 50/60 Hz motor with Schuko UK and US power leads |
| Dimensioner (L x B x H) | 44.5 x 15.0 x 23.5 cm (17.5 x 5.9 x 9.25 in.) |
| Watt | 90 w |
| Vægt (engelsk) | 47.3 lb. |
| Vægt (metrisk) | 21.5 kg |
| Flowhastighed | 120 to 143 L/min. |
| Til brug med | Labovac 10 |
| Frekvens | 50/60 Hz |
| Pumpetype | Rotary Vane Vacuum Pump |
| Vis mere |