Få mere at vide
Rosuvastatin tert-Butyl Ester, TRC
Artikelnummer. 30426105
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30426105 1 g
30420908 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30426105 Leverandør Toronto Research Chemicals Leverandørnr. TRCR7005401G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 355806-00-7
Molekylær formel C26 H36 F N3 O6 S
Molekylvægt (g/mol) 537.64
Synonym 6-Heptenoic acid, 7-[4-(4-fluorophenyl)-6-(1-methylethyl)-2-[methyl(methylsulfonyl)amino]-5-pyrimidinyl]-3,5-dihydroxy-, 1,1-dimethylethyl ester, (3R,5S,6E)-, (3R,5S,6E)-7-[4-(4-Fluorophenyl)-6-isopropyl-2-[(methanesulfonyl)methylamino]pyrimidin-5-yl]-3,5-dihydroxyhept-6-enoic acid tert-butyl ester, Rosuvastatin tert-butyl ester, tert-Butyl (6E)-7-[4-(4-fluorophenyl)-6-isopropyl-2-[methyl(methylsulfonyl)amino]pyrimidin-5-yl]-(3R,5S)-3,5-dihydroxyhept-6-enoate, tert-Butyl rosuvastatin, Rosuvastatin tert-Butyl Ester
IUPAC navn tert-butyl (E,3R,5S)-7-[4-(4-fluorophenyl)-2-[methyl(methylsulfonyl)amino]-6-propan-2-ylpyrimidin-5-yl]-3,5-dihydroxyhept-6-enoate
SMIL CC(C)c1nc(nc(c1\C=C\[C@@H](O)C[C@@H](O)CC(=O)OC(C)(C)C)c2ccc(F)cc2)N(C)S(=O)(=O)C
Tekniske data
Kemisk navn eller materiale Rosuvastatin tert-Butyl Ester
Mængde 1 g
InChI formel InChI=1S/C26H36FN3O6S/c1-16(2)23-21(13-12-19(31)14-20(32)15-22(33)36-26(3,4)5)24(17-8-10-18(27)11-9-17)29-25(28-23)30(6)37(7,34)35/h8-13,16,19-20,31-32H,14-15H2,1-7H3/b13-12+/t19-,20-/m1/s1
Formel vægt 537.2309