Få mere at vide
Rosuvastatin Methyl Ester, TRC
Artikelnummer. 17974915
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17974915 1 g
17934725 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17974915 Leverandør Toronto Research Chemicals Leverandørnr. TRCR7005651G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 147118-40-9
Molekylær formel C23 H30 F N3 O6 S
Molekylvægt (g/mol) 495.56
Synonym 6-Heptenoic acid, 7-[4-(4-fluorophenyl)-6-(1-methylethyl)-2-[methyl(methylsulfonyl)amino]-5-pyrimidinyl]-3,5-dihydroxy-, methyl ester, [S-[R*,S*-(E)]]-, 6-Heptenoic acid, 7-[4-(4-fluorophenyl)-6-(1-methylethyl)-2-[methyl(methylsulfonyl)amino]-5-pyrimidinyl]-3,5-dihydroxy-, methyl ester, (3R,5S,6E)-, Rosuvastatin methyl ester, Rosuvastatin Methyl Ester
IUPAC navn methyl (E,3R,5S)-7-[4-(4-fluorophenyl)-2-[methyl(methylsulfonyl)amino]-6-propan-2-ylpyrimidin-5-yl]-3,5-dihydroxyhept-6-enoate
SMIL COC(=O)C[C@H](O)C[C@H](O)\C=C\c1c(nc(nc1c2ccc(F)cc2)N(C)S(=O)(=O)C)C(C)C
Tekniske data
Kemisk navn eller materiale Rosuvastatin Methyl Ester
Mængde 1 g
InChI formel InChI=1S/C23H30FN3O6S/c1-14(2)21-19(11-10-17(28)12-18(29)13-20(30)33-4)22(15-6-8-16(24)9-7-15)26-23(25-21)27(3)34(5,31)32/h6-11,14,17-18,28-29H,12-13H2,1-5H3/b11-10+/t17-,18-/m1/s1
Formel vægt 495.1839