Få mere at vide
Rosuvastatin Methyl Ester, Mikromol™
Artikelnummer. 17921546
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17921546 25 mg
17913536 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17921546 Leverandør Mikromol Leverandørnr. MM0936.060025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Rosuvastatin Methyl Ester
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Rosuvastatin Methyl Ester
CAS 147118-40-9
Analyt- eller komponentnavne Rosuvastatin Methyl Ester
Forsendelsestilstand Room Temperature
Molekylær formel C23 H30 F N3 O6 S
Certificeringer/compliance ISO 17025
Synonym 6-Heptenoic acid, 7-[4-(4-fluorophenyl)-6-(1-methylethyl)-2-[methyl(methylsulfonyl)amino]-5-pyrimidinyl]-3,5-dihydroxy-, methyl ester, [S-[R*,S*-(E)]]-, 6-Heptenoic acid, 7-[4-(4-fluorophenyl)-6-(1-methylethyl)-2-[methyl(methylsulfonyl)amino]-5-pyrimidinyl]-3,5-dihydroxy-, methyl ester, (3R,5S,6E)-, Rosuvastatin methyl ester, Rosuvastatin Methyl Ester
InChI formel InChI=1S/C23H30FN3O6S/c1-14(2)21-19(11-10-17(28)12-18(29)13-20(30)33-4)22(15-6-8-16(24)9-7-15)26-23(25-21)27(3)34(5,31)32/h6-11,14,17-18,28-29H,12-13H2,1-5H3/b11-10+/t17-,18-/m1/s1
SMIL COC(=O)C[C@H](O)C[C@H](O)\C=C\c1c(nc(nc1c2ccc(F)cc2)N(C)S(=O)(=O)C)C(C)C
IUPAC navn methyl (E,3R,5S)-7-[4-(4-fluorophenyl)-2-[methyl(methylsulfonyl)amino]-6-propan-2-ylpyrimidin-5-yl]-3,5-dihydroxyhept-6-enoate
Molekylvægt (g/mol) 495.56
Mængde 25 mg
Formel vægt 495.1839 g/mol