missing translation for 'onlineSavingsMsg'
Learn More

Rosuvastatin Ethyl Ester, Mikromol™

Product Code. 17911776
Change view
Click to view available options
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
17911776 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17911776 Supplier Mikromol Supplier No. MM0936.380025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Rosuvastatin Ethyl Ester
TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Rosuvastatin Ethyl Ester
CAS 851443-04-4
Analyt- eller komponentnavne Rosuvastatin Ethyl Ester
Forsendelsestilstand Room Temperature
Molekylær formel C24 H32 F N3 O6 S
Certificeringer/compliance ISO 9001
Synonym (3R,5S,6E)-7-[4-(4-Fluorophenyl)-6-(1-methylethyl)-2-[methyl(methylsulfonyl)amino]-5-pyrimidinyl]-3,5-dihydroxy-6-heptenoic Acid Ethyl Ester, Rosuvastatin Ethyl Ester
InChI formel InChI=1S/C24H32FN3O6S/c1-6-34-21(31)14-19(30)13-18(29)11-12-20-22(15(2)3)26-24(28(4)35(5,32)33)27-23(20)16-7-9-17(25)10-8-16/h7-12,15,18-19,29-30H,6,13-14H2,1-5H3/b12-11+/t18-,19-/m1/s1
SMIL CCOC(=O)C[C@H](O)C[C@H](O)\C=C\c1c(nc(nc1c2ccc(F)cc2)N(C)S(=O)(=O)C)C(C)C
IUPAC navn ethyl (E,3R,5S)-7-[4-(4-fluorophenyl)-2-[methyl(methylsulfonyl)amino]-6-propan-2-ylpyrimidin-5-yl]-3,5-dihydroxyhept-6-enoate
Molekylvægt (g/mol) 509.59
Mængde 25 mg
Formel vægt 509.1996 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.