Få mere at vide
Rosuvastatin-d3 Sodium Salt, TRC
Artikelnummer. 30421873
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30421873 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30421873 Leverandør Toronto Research Chemicals Leverandørnr. TRCR7005011MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Rosuvastatin-d3 Sodium Salt
CAS 1279031-70-7
Renhedsgrad noter HPLC
Molekylær formel C22H24D3FN3NaO6S
Mængde 1 mg
Synonym (3R,5S,6E)-7-[4-(4-Fluorophenyl)-6-(1-methylethyl)-2-[methyl(methylsulfonyl-d3)amino]-5-pyrimidinyl]-3,5-dihydroxy-6-heptenoic Acid Monosodium Salt, [S-[R*,S*-(E)]]-7-[4-(4-Fluorophenyl)-6-(1-methylethyl)-2-[methyl(methylsulfonyl-d3)amino]-5-pyrimidinyl]-3,5-dihydroxy-6-heptenoic Acid Monosodium Salt, (3R,5S,6E)-7-[4-(4-Fluorophenyl)-6-isopropyl-2-[(methanesulfonyl-d3)methylamino]pyrimidin-5-yl]-3,5-dihydroxyhept-6-enoic Acid Sodium Salt, Rosuvastatin-d3 Sodium
InChI formel InChI=1S/C22H28FN3O6S.Na/c1-13(2)20-18(10-9-16(27)11-17(28)12-19(29)30)21(14-5-7-15(23)8-6-14)25-22(24-20)26(3)33(4,31)32;/h5-10,13,16-17,27-28H,11-12H2,1-4H3,(H,29,30);/q;+1/p-1/b10-9+;/t16-,17-;/m1./s1/i3D3;
SMIL [Na+].[2H]C([2H])([2H])N(c1nc(C(C)C)c(\C=C\[C@@H](O)C[C@@H](O)CC(=O)[O-])c(n1)c2ccc(F)cc2)S(=O)(=O)C
Molekylvægt (g/mol) 506.54
Procent renhed >95