missing translation for 'onlineSavingsMsg'
Få mere at vide

Rosuvastatin Calcium, Mikromol™

Artikelnummer. 18061970
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18061970 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Artikelnummer. 18061970 Leverandør Mikromol Leverandørnr. MM0936.00

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Rosuvastatin Calcium
TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Rosuvastatin Calcium
CAS 147098-20-2
Analyt- eller komponentnavne Rosuvastatin Calcium
Forsendelsestilstand Room Temperature
Molekylær formel 2 C22 H27 F N3 O6 S . Ca
Certificeringer/compliance ISO 17025
Synonym 6-Heptenoic acid, 7-[4-(4-fluorophenyl)-6-(1-methylethyl)-2-[methyl(methylsulfonyl)amino]-5-pyrimidinyl]-3,5-dihydroxy-, calcium salt (2:1), (3R,5S,6E)-, 6-Heptenoic acid, 7-[4-(4-fluorophenyl)-6-(1-methylethyl)-2-[methyl(methylsulfonyl)amino]-5-pyrimidinyl]-3,5-dihydroxy-, calcium salt (2:1), [S-[R*,S*-(E)]]-, Crestomed, Crestor, Fortius, Rostar, Rosuvastatin calcium, Rosuvastatin calcium salt, Rosuvastatin hemicalcium, Rozavel, S 4522, Suvikan, ZD 4522, calcium salt
InChI formel InChI=1S/2C22H28FN3O6S.Ca/c2*1-13(2)20-18(10-9-16(27)11-17(28)12-19(29)30)21(14-5-7-15(23)8-6-14)25-22(24-20)26(3)33(4,31)32;/h2*5-10,13,16-17,27-28H,11-12H2,1-4H3,(H,29,30);/q;;+2/p-2/b2*10-9+;/t2*16-,17-;/m11./s1
SMIL [Ca+2].CC(C)c1nc(nc(c1\C=C\[C@@H](O)C[C@@H](O)CC(=O)[O-])c2ccc(F)cc2)N(C)S(=O)(=O)C.CC(C)c3nc(nc(c3\C=C\[C@@H](O)C[C@@H](O)CC(=O)[O-])c4ccc(F)cc4)N(C)S(=O)(=O)C
IUPAC navn calcium;(E,3R,5S)-7-[4-(4-fluorophenyl)-2-[methyl(methylsulfonyl)amino]-6-propan-2-ylpyrimidin-5-yl]-3,5-dihydroxyhept-6-enoate
Molekylvægt (g/mol) 1001.14
Mængde 250 mg
Formel vægt 1000.2835 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.