Få mere at vide
Tocris Bioscience™ Rostafuroxin
GREENER_CHOICE
Na+/K+ ATPase modulator; antihypertensive agent
Brand:  Tocris Bioscience™ 5315/50
Encompass
Produktkode 15736696
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
Rostafuroxin is a Na+/K+ ATPase modulator. Inhibits the Normalizes renal Na+/K+ ATPase pump function in mutant adducin- and endogenous ouabain-dependent forms of hypertension. Reduces blood pressure in Milan hypertensive rats. Controls mania-like behaviors in Myshkin mice having a Na+/K+ ATPase α3 subunit mutation. Antagonizes the effects of Ouabain (Cat.No. 1076) on myogenic tone and Ca2+ signaling in rat hippocampal neurons.
Tekniske data
Tekniske data
3β,5β,14β)-21,23-Epoxy-24-norchola-20,22-diene-3,14,17-triol
156722-18-8
PST 2238
50 mg
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner