Få mere at vide
Tocris Bioscience™ Rosiglitazone
Potent and selective PPARγ agonist; antidiabetic agent
Brand: Tocris Bioscience™ 5325/10
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Rosiglitazone is a potent and selective PPARγ agonist (EC50 = 60 nM); exhibits no activity at PPARα and PPARβ. Promotes differentiation of pluripotent C3H10T1/2 stem cells into adipocytes. Also promotes differentiation of urothelial organoids in combination with erlotinib. Exhibits antihyperglycemic activity in diabetic ob/ob mouse model. Antidiabetic agent.Tekniske data
| 5-[[4-[2-(Methyl-2-pyridinylamino)ethoxy]phenyl]methyl]-2,4-thiazolidinedione | |
| 122320-73-4 | |
| BRL 49653 | |
| 10 mg |