Få mere at vide
Rosiglitazone-d3, TRC
Artikelnummer. 30426534
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30426534 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30426534 Leverandør Toronto Research Chemicals Leverandørnr. TRCR6935021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Rosiglitazone-D3
CAS 1132641-22-5
Molekylær formel C18 2H3 H16 N3 O3 S
Mængde 1 mg
Synonym 5-[[4-[2-(Methyl-d3-2-pyridinylamino)ethoxy]phenyl]methyl]-2,4-thiazolidinedione
InChI formel InChI=1S/C18H19N3O3S/c1-21(16-4-2-3-9-19-16)10-11-24-14-7-5-13(6-8-14)12-15-17(22)20-18(23)25-15/h2-9,15H,10-12H2,1H3,(H,20,22,23)/i1D3
SMIL [2H]C([2H])([2H])N(CCOc1ccc(CC2SC(=O)NC2=O)cc1)c3ccccn3
IUPAC navn 5-[[4-[2-[pyridin-2-yl(trideuteriomethyl)amino]ethoxy]phenyl]methyl]-1,3-thiazolidine-2,4-dione
Molekylvægt (g/mol) 360.45
Formel vægt 360.1335