Få mere at vide
Ropivacaine Imidazol, TRC
Artikelnummer. 30011384
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30011384 10 mg
30007693 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30011384 Leverandør Toronto Research Chemicals Leverandørnr. TRCR37990510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1945965-95-6
Molekylær formel C17 H24 N2 O
Molekylvægt (g/mol) 272.38
Synonym (8AS)-2-(2,6-dimethylphenyl)-3,3-dimethyl-6,7,8,8a-tetrahydro-5H-imidazo[1,5-a]pyridin-1-one
IUPAC navn (8aS)-2-(2,6-dimethylphenyl)-3,3-dimethyl-6,7,8,8a-tetrahydro-5H-imidazo[1,5-a]pyridin-1-one
SMIL Cc1cccc(C)c1N2C(=O)[C@@H]3CCCCN3C2(C)C
Tekniske data
Kemisk navn eller materiale Ropivacaine Imidazol
Mængde 10 mg
InChI formel InChI=1S/C17H24N2O/c1-12-8-7-9-13(2)15(12)19-16(20)14-10-5-6-11-18(14)17(19,3)4/h7-9,14H,5-6,10-11H2,1-4H3/t14-/m0/s1
Formel vægt 272.1889