Få mere at vide
Ropivacaine Hydrochloride Monohydrate, TRC
Artikelnummer. 30138761
Skift visning
Mængde:
10 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138761 250 mg
30005142 50 mg
30005559 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30138761 Leverandør Toronto Research Chemicals Leverandørnr. TRCR675000250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 132112-35-7
Tekniske data
Kemisk navn eller materiale Ropivacaine hydrochloride monohydrate
Renhedsgrad noter HPLC
Molekylær formel C17 H26 N2 O . Cl H . H2 O
Mængde 250 mg
Synonym Ropivacaine hydrochloride monohydrate, 2-Piperidinecarboxamide, N-(2,6-dimethylphenyl)-1-propyl-, hydrochloride, hydrate (1:1:1), (2S)-, 2-Piperidinecarboxamide, N-(2,6-dimethylphenyl)-1-propyl-, monohydrochloride, monohydrate, (2S)- (9CI), 2-Piperidinecarboxamide, N-(2,6-dimethylphenyl)-1-propyl-, monohydrochloride, monohydrate, (S)-, (2S)-2-[(Dimethylphenyl)carbamoyl]-1-propylpiperidin-1-ium chloride monohydrate, (-)-(2S)-N-(2,6-Dimethylphenyl)-1-propylpiperidine-2-carboxamide hydrochloride monohydrate
InChI formel InChI=1S/C17H26N2O.ClH.H2O/c1-4-11-19-12-6-5-10-15(19)17(20)18-16-13(2)8-7-9-14(16)3;;/h7-9,15H,4-6,10-12H2,1-3H3,(H,18,20);1H;1H2/t15-;;/m0../s1
SMIL O.Cl.CCCN1CCCC[C@H]1C(=O)Nc2c(C)cccc2C
IUPAC navn (2S)-N-(2,6-dimethylphenyl)-1-propylpiperidine-2-carboxamide;hydrate;hydrochloride
Molekylvægt (g/mol) 328.88
Formel vægt 328.1918
Procent renhed >95%