Få mere at vide
(±)-Ropivacaine-d7 (propyl-d7), CDN
Artikelnummer. 30317499
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30317499 0.05 g
30320323 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30317499 Leverandør CDN Leverandørnr. CDND69340.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (+/-)-Ropivacaine-D7 (N-propyl-D7)
CAS 1392208-04-6
Molekylær formel C17 D7 H19 N2 O
Mængde 0.05 g
Synonym N-(2,6-Dimethylphenyl)-1-(propyl-d7)-2-piperidinecarboxamide, (±)-Ropivacaine-d7, rac-Ropivacaine-d7, (±)-Ropivacaine-D7 (N-propyl-D7)
InChI formel InChI=1 S/C17H26N2O/c1-4-11-19-12-6-5-10-15(19)17(20)18-16-13(2)8-7-9-14(16)3/h7-9,15 H,4-6,10-12H2,1-3H3,(H,18,20)/i1D3,4D2,11D2
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])N1CCCCC1C(=O)Nc2c(C)cccc2C
IUPAC navn N-(2,6-dimethylphenyl)-1-(1,1,2,2,3,3,3-heptadeuteriopropyl)piperidine-2-carboxamide
Molekylvægt (g/mol) 281.444
Formel vægt 281.248 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity