Få mere at vide
Ropivacaine-d7 Hydrochloride, TRC
Artikelnummer. 30427038
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30427038 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30427038 Leverandør Toronto Research Chemicals Leverandørnr. TRCR6750021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ropivacaine-d7 Hydrochloride
CAS 1217667-10-1
Renhedsgrad noter HPLC
Molekylær formel C17H20D7ClN2O
Mængde 1 mg
Synonym (2S)-N-(2,6-Dimethylphenyl)-1-(1,1,2,2,3,3,3-heptadeuteriopropyl)piperidine-2-carboxamide hydrochloride, (2S)-N-(2,6-Dimethylphenyl)-1-(propyl-d7)-2-piperidinecarboxamide hydrochloride, Ropivacaine-d7 hydrochloride, Ropivacaine-D7 (N-propyl-D7) hydrochloride
InChI formel InChI=1S/C17H26N2O.ClH/c1-4-11-19-12-6-5-10-15(19)17(20)18-16-13(2)8-7-9-14(16)3;/h7-9,15H,4-6,10-12H2,1-3H3,(H,18,20);1H/t15-;/m0./s1/i1D3,4D2,11D2;
SMIL Cl.[2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])N1CCCC[C@H]1C(=O)Nc2c(C)cccc2C
Molekylvægt (g/mol) 317.91
Formel vægt 317.2251
Procent renhed >95