Få mere at vide
Roflumilast, TRC
Artikelnummer. 30012845
Skift visning
Mængde:
5 mg
10 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30012845 1 g
17931525 5 mg
17922645 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30012845 Leverandør Toronto Research Chemicals Leverandørnr. TRCR6397001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 162401-32-3
Molekylær formel C17 H14 Cl2 F2 N2 O3
Molekylvægt (g/mol) 403.21
Synonym Benzamide, 3-(cyclopropylmethoxy)-N-(3,5-dichloro-4-pyridinyl)-4-(difluoromethoxy)-, 3-(Cyclopropylmethoxy)-N-(3,5-dichloro-4-pyridinyl)-4-(difluoromethoxy)benzamide, ARQ 151, B 9302-107, BY 217, BYK 20869, Daliresp, Daxas, Libertek, Roflumilast
IUPAC navn 3-(cyclopropylmethoxy)-N-(3,5-dichloropyridin-4-yl)-4-(difluoromethoxy)benzamide
SMIL FC(F)Oc1ccc(cc1OCC2CC2)C(=O)Nc3c(Cl)cncc3Cl
Tekniske data
Kemisk navn eller materiale Roflumilast
Mængde 1 g
InChI formel InChI=1S/C17H14Cl2F2N2O3/c18-11-6-22-7-12(19)15(11)23-16(24)10-3-4-13(26-17(20)21)14(5-10)25-8-9-1-2-9/h3-7,9,17H,1-2,8H2,(H,22,23,24)
Formel vægt 402.035