missing translation for 'onlineSavingsMsg'
Learn More
Roflumilast N-oxide, MedChemExpress Product Code.: 16483108

Roflumilast N-oxide, MedChemExpress

Product Code. p-200099617
10 mg, 10mg
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16483108

Brand: Medchem Express HY100639

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Roflumilast N-oxide is a PDE type 4 inhibitor.

Roflumilast N-oxide is a PDE type 4 inhibitor.

Specifications

Chemical Name or Material Roflumilast N-oxide
CAS 292135-78-5
Color White
Purity Grade Notes Research
Molecular Formula C17H14Cl2F2N2O4
Quantity 10 mg
Solubility Information DMSO : 250 mg/mL (596.36 mM; Need ultrasonic)
SMILES O=C(NC1=C(Cl)C=N(C=C1Cl)=O)C2=CC=C(OC(F)F)C(OCC3CC3)=C2
Molecular Weight (g/mol) 419.21
Formula Weight 419.21
Percent Purity 99.52%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Roflumilast N-oxide, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.