Få mere at vide
Roflumilast-d4, TRC
Artikelnummer. 30420441
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30420441 10 mg
30415386 1 mg
30414923 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30420441 Leverandør Toronto Research Chemicals Leverandørnr. TRCR63970210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1398065-69-4
Molekylær formel C17 D4 H10 Cl2 F2 N2 O3
Molekylvægt (g/mol) 407.232
Synonym N-(3,5-Dichloropyridin-4-yl)-4-(difluoromethoxy)-3-[(2,2,3,3-tetradeuteriocyclopropyl)methoxy]benzamide, 3-[(Cyclopropyl-2,2,3,3-d4)methoxy]-N-(3,5-dichloro-4-pyridinyl)-4-(difluoromethoxy)benzamide, Roflumilast-d4, Roflumilast-D4 (cyclopropyl-2,2,3,3-D4)
IUPAC navn N-(3,5-dichloropyridin-4-yl)-4-(difluoromethoxy)-3-[(2,2,3,3-tetradeuteriocyclopropyl)methoxy]benzamide
SMIL [2H]C1([2H])C(COc2cc(ccc2OC(F)F)C(=O)Nc3c(Cl)cncc3Cl)C1([2H])[2H]
Tekniske data
Kemisk navn eller materiale Roflumilast-d4
Renhedsgrad noter HPLC
Mængde 10 mg
InChI formel InChI=1S/C17H14Cl2F2N2O3/c18-11-6-22-7-12(19)15(11)23-16(24)10-3-4-13(26-17(20)21)14(5-10)25-8-9-1-2-9/h3-7,9,17H,1-2,8H2,(H,22,23,24)/i1D2,2D2
Formel vægt 406.06
Procent renhed >95%