Få mere at vide
Roflumilast-d4 N-Oxide, TRC
Artikelnummer. 30416994
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30416994 10 mg
30414117 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30416994 Leverandør Toronto Research Chemicals Leverandørnr. TRCR63971210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1794760-31-8
Molekylær formel C17H10D4Cl2F2N2O4
Molekylvægt (g/mol) 423.23
Synonym 3-(Cyclopropylmethoxy-d4)-N-(3,5-dichloro-1-oxido-4-pyridinyl)-4-(difluoromethoxy)benzamide, N-(3,5-Dichloro-1-oxopyridin-4-yl)-4-difluoromethoxy-3-(cyclopropyl-d4) methoxybenzamide
IUPAC navn N-(3,5-dichloro-1-oxopyridin-4-yl)-4-(difluoromethoxy)-3-[(2,2,3,3-tetradeuteriocyclopropyl)methoxy]benzamide
SMIL ClC(C(NC(C1=CC(OCC2C([2H])([2H])C2([2H])[2H])=C(OC(F)F)C=C1)=O)=C(Cl)C=3)=CN3=O
Tekniske data
Kemisk navn eller materiale Roflumilast-d4 N-Oxide
Mængde 10 mg
InChI formel InChI=1S/C17H14Cl2F2N2O4/c18-11-6-23(25)7-12(19)15(11)22-16(24)10-3-4-13(27-17(20)21)14(5-10)26-8-9-1-2-9/h3-7,9,17H,1-2,8H2,(H,22,24)/i1D2,2D2
Formel vægt 422.055