Learn More
Thermo Scientific Chemicals Rocuronium bromide, 97%
Product Code. 16574454 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16574454 250 mg 250mg
16564454 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16574454 Supplier Thermo Scientific Chemicals Supplier No. 465022500
 Restricted Item
På lager
Add to basket
CAS: 119302-91-9 | C32H53BrN2O4 | 609.69 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 119302-91-9
InChI Key OYTJKRAYGYRUJK-FMCCZJBLSA-M
IUPAC Name 1-[(1R,2S,3aS,3bR,5aS,7S,8S,9aS,9bS,11aS)-1-(acetyloxy)-7-hydroxy-9a,11a-dimethyl-8-(morpholin-4-yl)-hexadecahydro-1H-cyclopenta[a]phenanthren-2-yl]-1-(prop-2-en-1-yl)pyrrolidin-1-ium bromide
SMILES [Br-].CC(=O)O[C@H]1[C@H](C[C@H]2[C@@H]3CC[C@H]4C[C@H](O)[C@H](C[C@]4(C)[C@H]3CC[C@]12C)N1CCOCC1)[N+]1(CC=C)CCCC1
Specifications
Melting Point 162.0°C to 164.0°C
Assay Percent Range 97%
Molecular Formula C32H53BrN2O4
Quantity 250 mg
Molecular Weight (g/mol) 609.69
Formula Weight 609.68
Chemical Name or Material Rocuronium bromide