missing translation for 'onlineSavingsMsg'
Få mere at vide

Robinetin, TRC

Artikelnummer. 30416507
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30416507 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30416507 Leverandør Toronto Research Chemicals Leverandørnr. TRCR6395155MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Robinetin
CAS 490-31-3
Molekylær formel C15 H10 O7
Mængde 5 mg
Synonym 3,7-Dihydroxy-2-(3,4,5-trihydroxyphenyl)-4H-1-benzopyran-1-one, 3,3',4',5',7-Pentahydroxyflavone, Norkanugin
InChI formel InChI=1S/C15H10O7/c16-7-1-2-8-11(5-7)22-15(14(21)12(8)19)6-3-9(17)13(20)10(18)4-6/h1-5,16-18,20-21H
SMIL OC1=C(Oc2cc(O)ccc2C1=O)c3cc(O)c(O)c(O)c3
IUPAC navn 3,7-dihydroxy-2-(3,4,5-trihydroxyphenyl)chromen-4-one
Molekylvægt (g/mol) 302.24
Formel vægt 302.0427
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.