Få mere at vide
Robenidine-d8, TRC
Artikelnummer. 30412750
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30412750 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30412750 Leverandør Toronto Research Chemicals Leverandørnr. TRCR6390021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Robenidine-d8
CAS 1173149-51-3
Molekylær formel C15H5D8Cl2N5
Mængde 1 mg
Synonym 2,2'-Bis[(4-chlorophenyl)methylene]carbonimidic Dihydrazide-d8, Bis[(4-chlorophenyl)methylene]carbonimidic Dihydrazide-d8, 1,3-Bis[(p-chlorobenzylidene)amino]guanidine-d8, 1,3-Bis(4-chlorobenzylideneamino) guanidine-d8, 1,3-Bis(p-chlorobenzylideneamino)guanidine-d8, Chimcoccid-d8, Khimcoccid-d8
InChI formel InChI=1S/C15H13Cl2N5/c16-13-5-1-11(2-6-13)9-19-21-15(18)22-20-10-12-3-7-14(17)8-4-12/h1-10H,(H3,18,21,22)/b19-9+,20-10+
SMIL [2H]C1=C(C(=C(C(=C1/C=N/N/C(=N/N=C/C2=C(C(=C(C(=C2[2H])[2H])Cl)[2H])[2H])/N)[2H])[2H])Cl)[2H]
Molekylvægt (g/mol) 342.25