missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ RNF141 Recombinant Protein

2735.00 DKK - 4145.00 DKK

Specifications

Accession Number AAH18104
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Gene ID (Entrez) 50862
Molecular Weight (g/mol) 50.93
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16169462
View Documents
Abnova™
H00050862-P01.10ug
10 ÎĽg
2735.00 DKK
10µg
Please to purchase this item. Need a web account? Register with us today!
16179462
View Documents
Abnova™
H00050862-P01.25ug
25 ÎĽg
4145.00 DKK
25µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications
Show More
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Abnova™ RNF141 Recombinant Protein

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.