missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human RNAse H2A Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP0787
Additional Details : Weight : 0.01000kg
Description
RNAse H2A Synthetic Peptide, PEP-0787, from Invitrogen.
This peptide corresponds to 17 amino acids near the center of human RNAse H2A.PEP-0787 can be used as a blocking peptide with polyclonal antibody PA5-20667.Specifications
10535 | |
purified | |
Human RNAse H2A | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
O75792 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
RNASEH2A | |
0.02% sodium azide | |
50 μg | |
PBS |