Learn More
Rnase-free Water, non DEPCAbnova™
Product Code. 16032048
Change view
Click to view available options
Quantity:
5 x 1 mL
2 x 50 mL
500 mL
Unit Size:
500mL
Pack of 2
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16032048 5 x 1 mL Pack of 5
16012048 2 x 50 mL Pack of 2
16022048 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16032048 Supplier Abnova Supplier No. R0013.5x1mL
 Restricted Item
Forventet forsendelse: 01-04-2027
RNase-free Water is suitable to experiment and research of RNA.
  • The RNase-free Water, non DEPC was prepared from highly purified water by ultrafiltration without DEPC treatment.
  • The product was confirmed to be free from RNase and DNase.
  • Apperance is clear liquid.
Specifications
Quantity 5 x 1 mL