Learn More
Thermo Scientific™ RNA sample loading buffer (6X)
Brand:  Thermo Scientific™ J62468.S0
387.22 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15404849
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
6X
Liquid
2mL
Store at -20°C
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN14E502 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number
Lot NumberP10C568 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number
Lot NumberQ01H517 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot NumberQ24F513 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot NumberN14E501 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ RNA sample loading buffer (6X) > 2mL