Learn More
Cytiva RNA Homopolymers
Product Code. 11585115 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Product Code. Quantity Unit Size
11585115 500 mg 500mg
1 options
Product Code. 11585115 Supplier Cytiva Supplier No. 27411002
  • ONLINE EXCLUSIVE PRICE
    10150.00 DKK
    9508.00 DKK / 500mg
    Spare 642.00 DKK
    6% Slukket
 Restricted Item
Forventet forsendelse: 07-12-2026
Template primers
These products are dispensed by weight of dry material, which includes polymer, residual salt, and residual water; content of polymer may vary from lot to lot.
TRUSTED_SUSTAINABILITY
Specifications
Product Type RNA Homopolymer
Content And Storage -20°C
Primer Poly(rA)
Quantity 500 mg