Få mere at vide
Tocris Bioscience™ RN 9893 hydrochloride
Potent and selective TRPV4 antagonist
Brand: Tocris Bioscience™ 5678/50
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
RN 9893 hydrochloride is a potent and selective TRPV4 antagonist (IC50 values are 320, 420 and 660 nM for mouse, human and rat channels, respectively). Exhibits >15-fold selectivity for TRPV4 over TRPV1, TRPV3 and TRPM8, and a panel of 54 other common biological targets. Orally bioavailable.Tekniske data
| N-[4-[[4-(1-Methylethyl)-1-piperazinyl]sulfonyl]phenyl]-2-nitro-4-(trifluoromethyl)benzamide hydrochloride | |
| 2109450-40-8 | |
| 50 mg |