Få mere at vide
Rizatriptan Benzoate, LoGiCal
Artikelnummer. 30464144
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30464144 10 mg
1 options
Artikelnummer. 30464144 Leverandør LoGiCal Leverandørnr. LGCFOR1313.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Rizatriptan benzoate
CAS 145202-66-0
Analyt- eller komponentnavne Rizatriptan benzoate
Forsendelsestilstand Room Temperature
Molekylær formel C15 H19 N5 . C7 H6 O2
Certificeringer/compliance ISO 9001
Synonym Rizatriptan benzoate, N,N-Dimethyl-2-[5-(1H-1,2,4-triazol-1-ylmethyl)-1H-indol-3-yl]ethanamine benzoate, 1H-Indole-3-ethanamine, N,N-dimethyl-5-(1H-1,2,4-triazol-1-ylmethyl)-, benzoate (1:1), 1H-Indole-3-ethanamine, N,N-dimethyl-5-(1H-1,2,4-triazol-1-ylmethyl)-, monobenzoate (9CI), MK 462, Maxalt, Rizora
InChI formel InChI=1S/C15H19N5.C7H6O2/c1-19(2)6-5-13-8-17-15-4-3-12(7-14(13)15)9-20-11-16-10-18-20;8-7(9)6-4-2-1-3-5-6/h3-4,7-8,10-11,17H,5-6,9H2,1-2H3;1-5H,(H,8,9)
Løsningstype Neat
SMIL CN(C)CCc1c[nH]c2ccc(Cn3cncn3)cc12.OC(=O)c4ccccc4
IUPAC navn benzoic acid;N,N-dimethyl-2-[5-(1,2,4-triazol-1-ylmethyl)-1H-indol-3-yl]ethanamine
Industry Type Clinical Testing
Molekylvægt (g/mol) 391.47
Mængde 10 mg
Formel vægt 391.2008 g/mol