Få mere at vide
Rivastigmine, Dr. Ehrenstorfer
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Rivastigmine |
| CAS | 123441-03-2 |
| Analyt- eller komponentnavne | Rivastigmine |
| Forsendelsestilstand | +20°C |
| Molekylær formel | C14 H22 N2 O2 |
| Certificeringer/compliance | ISO/IEC 17025 |
| Synonym | 3-[(1S)-1-(Dimethylamino)ethyl]phenyl ethyl(methyl)carbamate, Carbamic acid, N-ethyl-N-methyl-, 3-[(1S)-1-(dimethylamino)ethyl]phenyl ester, Carbamic acid, ethylmethyl-, 3-[(1S)-1-(dimethylamino)ethyl]phenyl ester (9CI), Carbamic acid, ethylmethyl-, 3-[1-(dimethylamino)ethyl]phenyl ester, (S)-, ENA 713 free base, Exelon, Prometax, Rivastach, S-Rivastigmine, SDZ 212-713, [3-[(1S)-1-(Dimethylamino)ethyl]phenyl] N-ethyl-N-methylcarbamate |
| InChI formel | InChI=1S/C14H22N2O2/c1-6-16(5)14(17)18-13-9-7-8-12(10-13)11(2)15(3)4/h7-11H,6H2,1-5H3/t11-/m0/s1 |
| SMIL | CCN(C)C(=O)Oc1cccc(c1)[C@H](C)N(C)C |
| IUPAC navn | [3-[(1S)-1-(dimethylamino)ethyl]phenyl] N-ethyl-N-methylcarbamate |
| Vis mere |