Få mere at vide
Rivaroxaban N-Hydrolyse N-5-Chlorothiophene-2-carbaldehyde, TRC
Artikelnummer. 30005240
Skift visning
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30005240 5 mg
30008052 25 mg
30004609 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30005240 Leverandør Toronto Research Chemicals Leverandørnr. TRCR5380255MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1151893-81-0
Molekylær formel C24 H21 Cl2 N3 O7 S2
Molekylvægt (g/mol) 598.48
Synonym 2-[2-[[(5-Chloro-2-thienyl)carbonyl][4-[(5S)-5-[[[(5-chloro-2-thienyl)carbonyl]amino]methyl]-2-oxo-3-oxazolidinyl]phenyl]amino]ethoxy]acetic Acid, Rivaroxaban Amine Impurity
IUPAC navn 2-[2-[N-(5-chlorothiophene-2-carbonyl)-4-[(5S)-5-[[(5-chlorothiophene-2-carbonyl)amino]methyl]-2-oxo-1,3-oxazolidin-3-yl]anilino]ethoxy]acetic acid
SMIL OC(=O)COCCN(C(=O)c1ccc(Cl)s1)c2ccc(cc2)N3C[C@H](CNC(=O)c4ccc(Cl)s4)OC3=O
Tekniske data
Kemisk navn eller materiale Rivaroxaban N-Hydrolyse N-5-Chlorothiophene-2-carbaldehyde
Renhedsgrad noter HPLC
Mængde 5 mg
InChI formel InChI=1S/C24H21Cl2N3O7S2/c25-19-7-5-17(37-19)22(32)27-11-16-12-29(24(34)36-16)15-3-1-14(2-4-15)28(9-10-35-13-21(30)31)23(33)18-6-8-20(26)38-18/h1-8,16H,9-13H2,(H,27,32)(H,30,31)/t16-/m0/s1
Formel vægt 597.0198
Procent renhed >95%