Få mere at vide
Rivaroxaban-d4, TRC
Artikelnummer. 30426447
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30426447 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30426447 Leverandør Toronto Research Chemicals Leverandørnr. TRCR5380021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Rivaroxaban-d4
CAS 1132681-38-9
Renhedsgrad noter HPLC
Molekylær formel C19H14D4ClN3O5S
Mængde 1 mg
Synonym Rivaroxaban D4
InChI formel InChI=1S/C19H18ClN3O5S/c20-16-6-5-15(29-16)18(25)21-9-14-10-23(19(26)28-14)13-3-1-12(2-4-13)22-7-8-27-11-17(22)24/h1-6,14H,7-11H2,(H,21,25)/t14-/m0/s1/i1D,2D,3D,4D
SMIL [2H]c1c([2H])c(N2C[C@H](CNC(=O)c3ccc(Cl)s3)OC2=O)c([2H])c([2H])c1N4CCOCC4=O
IUPAC navn 5-chloro-N-[[(5S)-2-oxo-3-[2,3,5,6-tetradeuterio-4-(3-oxomorpholin-4-yl)phenyl]-1,3-oxazolidin-5-yl]methyl]thiophene-2-carboxamide
Molekylvægt (g/mol) 439.91
Formel vægt 439.0907
Procent renhed >95