Få mere at vide
Ritonavir-13C3, TRC
Artikelnummer. 30414053
Skift visning
Mængde:
0.5 mg
5 mg
Pakningsstørrelse:
0.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30414053 0.5 mg 0.5mg
30426104 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30414053 Leverandør Toronto Research Chemicals Leverandørnr. TRCR5350030.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1217673-23-8
Molekylær formel 13C3 C34 H48 N6 O5 S2
Molekylvægt (g/mol) 723.92
Synonym (3S,4S,6S,9S)-4-Hydroxy-12-methyl-9-(1-methylethyl)-13-[2-(1-methylethyl)-4-thiazolyl]-8,11-dioxo-3,6-bis(phenylmethyl)-2,7,10,12-tetraazatridecanoic Acid 5-Thiazolylmethyl-13C3 Ester, A 84538-13C3, ABT 538-13C3, Norvir-13C3
IUPAC navn (4,5-^{13}C_{2})thiazol-5-yl(1^{13}C)methyl N-[(1S,2S,4S)-1-benzyl-2-hydroxy-4-[[(2S)-2-[[(2-isopropylthiazol-4-yl)methyl-methyl-carbamoyl]amino]-3-methyl-butanoyl]amino]-5-phenyl-pentyl]carbamate
SMIL CC(C)[C@H](NC(=O)N(C)Cc1csc(n1)C(C)C)C(=O)N[C@H](C[C@H](O)[C@H](Cc2ccccc2)NC(=O)O[13CH2][13c]3[13cH]ncs3)Cc4ccccc4
Tekniske data
Kemisk navn eller materiale Ritonavir 4,5,methyl-methylthiazolyl-13C3
Mængde 0.5 mg
InChI formel InChI=1S/C37H48N6O5S2/c1-24(2)33(42-36(46)43(5)20-29-22-49-35(40-29)25(3)4)34(45)39-28(16-26-12-8-6-9-13-26)18-32(44)31(17-27-14-10-7-11-15-27)41-37(47)48-21-30-19-38-23-50-30/h6-15,19,22-25,28,31-33,44H,16-18,20-21H2,1-5H3,(H,39,45)(H,41,47)(H,42,46)/t28-,31-,32-,33-/m0/s1/i19+1,21+1,30+1
Formel vægt 723.3228