Få mere at vide
Ritodrine Hydrochloride, LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Ritodrine Hydrochloride |
| CAS | 23239-51-2 |
| Analyt- eller komponentnavne | Ritodrine Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C17 H21 N O3 . HCl |
| Certificeringer/compliance | ISO 9001 |
| Synonym | DU 21220, Miolene, NSC 291565, Pre-Par, Prempar, Ritodrine hydrochloride, Utemerin, Utopar, Yutopar, Benzenemethanol, 4-hydroxy-alpha-[(1R)-1-[[2-(4-hydroxyphenyl)ethyl]amino]ethyl]-, hydrochloride (1:1), (alphaS)-rel-, Benzenemethanol, 4-hydroxy-alpha-[(1R)-1-[[2-(4-hydroxyphenyl)ethyl]amino]ethyl]-, hydrochloride, (alphaS)-rel- (9CI), Benzenemethanol, 4-hydroxy-alpha-[1-[[2-(4-hydroxyphenyl)ethyl]amino]ethyl]-, hydrochloride, (R*,S*)-, Benzyl alcohol, p-hydroxy-alpha-[1-[(p-hydroxyphenethyl)amino]ethyl]-, hydrochloride, erythro- (8CI), (+/-)-Ritodrine hydrochloride |
| InChI formel | InChI=1S/C17H21NO3.ClH/c1-12(17(21)14-4-8-16(20)9-5-14)18-11-10-13-2-6-15(19)7-3-13;/h2-9,12,17-21H,10-11H2,1H3;1H/t12-,17-;/m1./s1 |
| Løsningstype | Neat |
| SMIL | Cl.C[C@@H](NCCc1ccc(O)cc1)[C@@H](O)c2ccc(O)cc2 |
| Vis mere |