Få mere at vide
Ritalinic-d9 Acid HCl (piperidine-d9) (mixture of stereoisomers), CDN
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Ritalinic Acid D9 (piperidine-D9) Hydrochloride |
| CAS | 1276197-13-7 |
| Molekylær formel | C13 D9 H8 N O2 . Cl H |
| Mængde | 0.01 g |
| Synonym | 2-(2,3,3,4,4,5,5,6,6-Nonadeuteriopiperidin-2-yl)-2-phenylacetic acid hydrochloride, α-Phenyl-2-(piperidine-d9)acetic acid hydrochloride, Ritalinic-d9 acid hydrochloride, Ritalinic acid-d9 hydrochloride, Ritalinic acid D9 (piperidine-D9) hydrochloride, 2-Phenyl-2-(piperidin-2,3,3,4,4,5,5,6,6-d9-2-yl)acetic acid hydrochloride, 2-Phenyl-2-(piperidin-2-yl-d9)acetic acid hydrochloride |
| InChI formel | InChI=1 S/C13H17NO2.ClH/c15-13(16)12(10-6-2-1-3-7-10)11-8-4-5-9-14-11;/h1-3,6-7,11-12,14 H,4-5,8-9H2,(H,15,16);1 H/i4D2,5D2,8D2,9D2,11 D; |
| SMIL | Cl.[2 H]C1([2 H])NC([2 H])(C(C(=O)O)c2ccccc2)C([2 H])([2 H])C([2 H])([2 H])C1([2 H])[2 H] |
| IUPAC navn | 2-(2,3,3,4,4,5,5,6,6-nonadeuteriopiperidin-2-yl)-2-phenylacetic acid;hydrochloride |
| Molekylvægt (g/mol) | 264.796 |
| Formel vægt | 264.159 g/mol |
| Vis mere |