Få mere at vide
Ritalinic Acid Hydrochloride, Mikromol™
Beskrivelse
Ritalinic Acid Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Ritalinic acid HCl |
| CAS | 19395-40-5 |
| Analyt- eller komponentnavne | Ritalinic acid HCl |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C13 H17 N O2 . HCl |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Ritalinic Acid Hydrochloride, Methylphenidate Related Compound A (USP), Methylphenidate Related Compound A as Hydrochloride |
| InChI formel | InChI=1S/C13H17NO2.ClH/c15-13(16)12(10-6-2-1-3-7-10)11-8-4-5-9-14-11;/h1-3,6-7,11-12,14H,4-5,8-9H2,(H,15,16);1H |
| SMIL | Cl.OC(=O)C(C1CCCCN1)c2ccccc2 |
| IUPAC navn | 2-phenyl-2-piperidin-2-ylacetic acid;hydrochloride |
| Vis mere |